Methyl 6-chloro-2-fluoro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

52860-5G 52860-25G
77365-136EA 131.84 USD
77365-136 77376-770
Methyl 6-chloro-2-fluoro-3-methylbenzoate ≥97%
Methyl 6-chloro-2-fluoro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD06203782
CAS Number: 773874-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR