N-(5-Fluoro-2-methylphenyl)pivalamide ≥97%

Supplier: AOBChem USA

Warning

52456-5G 52456-25G
77365-034EA 121.54 USD
77365-034 77376-664
N-(5-Fluoro-2-methylphenyl)pivalamide ≥97%
N-(5-Fluoro-2-methylphenyl)pivalamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆FNO
Storage Temperature: Ambient
MDL Number: MFCD08724331
CAS Number: 916037-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR