3-Chloro-[1,1'-biphenyl]-2-ol ≥97%

Supplier: AOBChem USA

Warning

52350-5G 52350-25G
77365-000EA 100.94 USD
77365-000 77376-616
3-Chloro-[1,1'-biphenyl]-2-ol ≥97%
3-Chloro-[1,1'-biphenyl]-2-ol

Precaución: for Research Use Only

Formula: C₁₂H₉ClO
MW: 204.652 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00152342
CAS Number: 85-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR