(4-Fluoro-3-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

52274-5G 52274-25G
77364-982EA 100.94 USD
77364-982 77376-592
(4-Fluoro-3-(trifluoromethyl)phenyl)(methyl)sulfane ≥97%
(4-Fluoro-3-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₆F₄S
MW: 210.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527246
CAS Number: 929212-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR