2,4-Dichlorobenzaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

52167-5G 52167-25G
77364-960EA 79.31 USD
77364-960 77376-556
2,4-Dichlorobenzaldehyde oxime ≥97%
2,4-Dichlorobenzaldehyde oxime

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO
Storage Temperature: Ambient
CAS Number: 56843-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR