N-(3,4-Dichlorophenyl)-2,2,2-trifluoroacetamide ≥97%

Supplier: AOBChem USA

Warning

51875-5G 51875-25G
77364-904EA 127.72 USD
77364-904 77376-436
N-(3,4-Dichlorophenyl)-2,2,2-trifluoroacetamide ≥97%
N-(3,4-Dichlorophenyl)-2,2,2-trifluoroacetamide

Precaución: for Research Use Only

Formula: C₈H₄Cl₂F₃NO
Storage Temperature: Ambient
MDL Number: MFCD00584859
CAS Number: 64694-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR