2,2,2-Trifluoro-N-(3-fluoro-4-methoxyphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

51547-5G 51547-25G
77364-780EA 131.84 USD
77364-780 77376-318
2,2,2-Trifluoro-N-(3-fluoro-4-methoxyphenyl)acetamide ≥97%
2,2,2-Trifluoro-N-(3-fluoro-4-methoxyphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₇F₄NO₂
MW: 237.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20148375
CAS Number: 1175563-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR