N-Cyclopropyl-2,4,6-trifluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

51393-5G 51393-25G
77364-736EA 138.02 USD
77364-736 77376-234
N-Cyclopropyl-2,4,6-trifluorobenzamide ≥97%
N-Cyclopropyl-2,4,6-trifluorobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₈F₃NO
MW: 215.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21477098
CAS Number: 1513248-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR