N'-((6-methoxypyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥95%

Supplier: AOBChem USA

Warning

50580-5G 50580-25G
77364-508EA 121.54 USD
77364-508 77376-008
N'-((6-methoxypyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide ≥95%
N'-((6-methoxypyridin-3-yl)methylene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₅N₃O₃S
MW: 305.352 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902671
CAS Number: 2586233-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR