Piperidin-1-yl(2,4,6-trichlorophenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

49838-5G 49838-25G
77364-410EA 166.86 USD
77364-410 77375-904
Piperidin-1-yl(2,4,6-trichlorophenyl)methanone ≥97%
Piperidin-1-yl(2,4,6-trichlorophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₂Cl₃NO
Storage Temperature: Ambient
MDL Number: MFCD34756647
CAS Number: 2764731-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR