2,5-Dichloro-N-isopropylbenzamide ≥97%

Supplier: AOBChem USA

Warning

49730-5G 49730-25G
77364-372EA 166.86 USD
77364-372 77375-852
2,5-Dichloro-N-isopropylbenzamide ≥97%
2,5-Dichloro-N-isopropylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₁Cl₂NO
MW: 232.106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01195573
CAS Number: 39887-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR