Ethyl dibenzo[b,d]thiophene-4-carboxylate ≥97%
Supplier: AOBChem USA
|
Warning
|
49679-5G
49679-25G
77364-336EA
166.86
USD
77364-336
77375-834
Ethyl dibenzo[b,d]thiophene-4-carboxylate ≥97%
Ethyl dibenzo[b,d]thiophene-4-carboxylate
Precaución: for Research Use Only
Formula:
C₁₅H₁₂O₂S MW: 256.32 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34178574 CAS Number: 2643368-19-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...