1,5-Dibromo-2-chloro-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

49588-5G 49588-25G
77364-314EA 100.94 USD
77364-314 77375-786
1,5-Dibromo-2-chloro-4-methylbenzene ≥97%
1,5-Dibromo-2-chloro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂Cl
MW: 284.376 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18089303
CAS Number: 150812-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR