N-(5-Chloro-2-methylphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

49257-5G 49257-25G
77364-236EA 114.33 USD
77364-236 77375-718
N-(5-Chloro-2-methylphenyl)acetamide ≥97%
N-(5-Chloro-2-methylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD00018743
CAS Number: 5900-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR