1,4-Dimethoxy-2-iodo-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

48870-5G 48870-25G
77364-064EA 124.63 USD
77364-064 77375-588
1,4-Dimethoxy-2-iodo-5-methylbenzene ≥97%
1,4-Dimethoxy-2-iodo-5-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO₂
Storage Temperature: Ambient
CAS Number: 75056-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR