N'-(3,5-Dibromobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47204-5G 47204-25G
77363-636EA 131.84 USD
77363-636 77375-188
N'-(3,5-Dibromobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(3,5-Dibromobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂Br₂N₂O₂S
MW: 432.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707242
CAS Number: 2432855-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR