N'-(5-Bromo-2-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47200-5G 47200-25G
77363-634EA 90.64 USD
77363-634 77375-186
N'-(5-Bromo-2-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(5-Bromo-2-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFN₂O₂S
MW: 371.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707241
CAS Number: 1303005-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR