N'-(2-Bromo-4-chlorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47192-5G 47192-25G
77363-632EA 166.86 USD
77363-632 77375-184
N'-(2-Bromo-4-chlorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(2-Bromo-4-chlorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrClN₂O₂S
MW: 387.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707240
CAS Number: 1821332-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR