N'-(4-Chloro-2-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47188-5G 47188-25G
77363-630EA 131.84 USD
77363-630 77375-182
N'-(4-Chloro-2-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(4-Chloro-2-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFN₂O₂S
MW: 326.774 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707239
CAS Number: 2432855-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR