N'-(4-Bromo-2-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47186-5G 47186-25G
77363-628EA 121.54 USD
77363-628 77375-180
N'-(4-Bromo-2-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(4-Bromo-2-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFN₂O₂S
MW: 371.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707238
CAS Number: 2432855-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR