N'-(2,3-Dichlorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47130-5G 47130-25G
77363-618EA 90.64 USD
77363-618 77375-170
N'-(2,3-Dichlorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(2,3-Dichlorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂Cl₂N₂O₂S
MW: 343.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707235
CAS Number: 347332-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR