N'-(2,3-Difluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47126-5G 47126-25G
77363-614EA 131.84 USD
77363-614 77375-166
N'-(2,3-Difluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(2,3-Difluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂N₂O₂S
MW: 310.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707233
CAS Number: 2432855-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR