N'-(5-Fluoro-2-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

47119-5G 47119-25G
77363-612EA 131.84 USD
77363-612 77375-164
N'-(5-Fluoro-2-methylbenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(5-Fluoro-2-methylbenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₅H₁₅FN₂O₂S
MW: 306.355 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707232
CAS Number: 2432855-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR