N'-(2-Bromo-4-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46992-5G 46992-25G
77363-594EA 166.86 USD
77363-594 77375-148
N'-(2-Bromo-4-fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(2-Bromo-4-fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFN₂O₂S
MW: 371.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707223
CAS Number: 2068051-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR