N'-(1-(4-Fluorophenyl)ethylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46586-5G 46586-25G
77363-510EA 160.68 USD
77363-510 77375-060
N'-(1-(4-Fluorophenyl)ethylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(1-(4-Fluorophenyl)ethylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₅H₁₅FN₂O₂S
MW: 306.355 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01353344
CAS Number: 75230-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR