N'-(4-Fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46570-5G 46570-25G
77363-506EA 166.86 USD
77363-506 77375-054
N'-(4-Fluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(4-Fluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₃FN₂O₂S
MW: 292.329 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00415867
CAS Number: 210422-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR