N'-(2,6-Difluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%

Supplier: AOBChem USA

Warning

46557-5G 46557-25G
77363-498EA 106.09 USD
77363-498 77375-046
N'-(2,6-Difluorobenzylidene)-4-methylbenzenesulfonohydrazide ≥97%
N'-(2,6-Difluorobenzylidene)-4-methylbenzenesulfonohydrazide

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂N₂O₂S
MW: 310.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707192
CAS Number: 2062613-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR