(4-Bromophenyl)(isobutyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

46295-5G 46295-25G
77363-414EA 80.34 USD
77363-414 77374-964
(4-Bromophenyl)(isobutyl)sulfane ≥97%
(4-Bromophenyl)(isobutyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrS
Storage Temperature: Ambient
MDL Number: MFCD20040742
CAS Number: 76542-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR