tert-Butyl 5-chloro-6-methoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

45422-5G 45422-25G
77363-212EA 131.84 USD
77363-212 77374-756
tert-Butyl 5-chloro-6-methoxynicotinate ≥97%
tert-Butyl 5-chloro-6-methoxynicotinate

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD34564077
CAS Number: 2733068-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR