Benzyl 2-bromobenzoate ≥97%

Supplier: AOBChem USA

Warning

39546-5G 39546-25G
77363-156EA 116.39 USD
77363-156 77374-700
Benzyl 2-bromobenzoate ≥97%
Benzyl 2-bromobenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD06460675
CAS Number: 67460-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR