4-Fluoro-2,3-dimethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

39348-5G 39348-25G
77363-154EA 173.04 USD
77363-154 77374-698
4-Fluoro-2,3-dimethylphenylboronic acid ≥97%
4-Fluoro-2,3-dimethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₂
MW: 167.973 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699619
CAS Number: 211495-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR