Benzyl 5-bromo-2-chloroisonicotinate ≥95%

Supplier: AOBChem USA

Warning

38736-5G 38736-25G
77363-130EA 90.64 USD
77363-130 77374-670
Benzyl 5-bromo-2-chloroisonicotinate ≥95%
Benzyl 5-bromo-2-chloroisonicotinate

Precaución: for Research Use Only

Formula: C₁₃H₉BrClNO₂
MW: 326.573 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707114
CAS Number: 1881980-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR