2,4,6-Trichloro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37961-5G 37961-25G
77362-840EA 162.74 USD
77362-840 77374-424
2,4,6-Trichloro-3-methoxybenzaldehyde ≥97%
2,4,6-Trichloro-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅Cl₃O₂
MW: 239.483 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902633
CAS Number: 704899-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR