8-Chloro[1,2,4]triazolo[4,3-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

36831-5G 36831-25G
77362-548EA 118.45 USD
77362-548 77374-134
8-Chloro[1,2,4]triazolo[4,3-a]pyridine ≥97%
8-Chloro[1,2,4]triazolo[4,3-a]pyridine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
Storage Temperature: Ambient
MDL Number: MFCD09258641
CAS Number: 501357-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR