4-Methoxy-3-(methoxymethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

36387-5G 36387-25G
77362-414EA 129.78 USD
77362-414 77374-008
4-Methoxy-3-(methoxymethyl)benzaldehyde ≥95%
4-Methoxy-3-(methoxymethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01822195
CAS Number: 76646-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR