3-Bromoveratrole ≥95%

Supplier: AOBChem USA

Warning

34686-5G 34686-25G
77362-050EA 76.22 USD
77362-050 77373-638
3-Bromoveratrole ≥95%
3-Bromoveratrole

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD00185638
CAS Number: 5424-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR