4-Bromo-2,6-diiodophenol ≥95%

Supplier: AOBChem USA

Warning

34436-5G 34436-25G
77362-014EA 79.31 USD
77362-014 77373-596
4-Bromo-2,6-diiodophenol ≥95%
4-Bromo-2,6-diiodophenol

Precaución: for Research Use Only

Formula: C₆H₃BrI₂O
Storage Temperature: Ambient
MDL Number: MFCD00672916
CAS Number: 15459-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR