4-Bromo-2,6-dichlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

34119-5G 34119-25G
77361-986EA 128.75 USD
77361-986 77373-564
4-Bromo-2,6-dichlorobenzoic acid ≥97%
4-Bromo-2,6-dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11227148
CAS Number: 232275-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR