3-Chloro-5-fluorosalicylaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-5-fluoro-2-hydroxybenzaldehyde

33545-5G 33545-25G
77361-896EA 119.48 USD
77361-896 77373-476
3-Chloro-5-fluorosalicylaldehyde ≥97%
3-Chloro-5-fluorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClFO₂
MW: 174.5569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039291
CAS Number: 82128-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR