3,6-Dibromobenzene-1,2-diamine ≥95%
Supplier: AOBChem USA
|
Warning
|
32199-5G
32199-25G
77361-686EA
131.84
USD
77361-686
77373-324
3,6-Dibromobenzene-1,2-diamine ≥95%
3,6-Dibromobenzene-1,2-diamine
Precaución: for Research Use Only
Formula:
C₆H₆Br₂N₂ MW: 265.93 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19288763 CAS Number: 69272-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...