5-Bromo-3-chloro-2-isobutoxypyridine ≥95%

Supplier: AOBChem USA

Warning

32133-5G 32133-25G
77361-682EA 128.75 USD
77361-682 77373-322
5-Bromo-3-chloro-2-isobutoxypyridine ≥95%
5-Bromo-3-chloro-2-isobutoxypyridine

Precaución: for Research Use Only

Formula: C₉H₁₁BrClNO
Storage Temperature: Ambient
MDL Number: MFCD22201175
CAS Number: 1289093-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR