3-Chloro-7-nitroquinoline ≥97%

Supplier: AOBChem USA

Warning

31640-5G 31640-25G
77361-642EA 152.44 USD
77361-642 77373-278
3-Chloro-7-nitroquinoline ≥97%
3-Chloro-7-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28339605
CAS Number: 75755-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR