4-Butoxy-2-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

31074-5G 31074-25G
77361-546EA 173.04 USD
77361-546 77373-224
4-Butoxy-2-fluorophenylboronic acid pinacol ester ≥97%
4-Butoxy-2-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₄BFO₃
Storage Temperature: Ambient
MDL Number: MFCD29088255
CAS Number: 2121514-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR