4-(3-Chloro-2-pyridinyl)morpholine ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Chloro-2-morpholinopyridine
30800-5G
30800-25G
77361-492EA
128.75
USD
77361-492
77373-188
4-(3-Chloro-2-pyridinyl)morpholine ≥95%
4-(3-Chloro-2-pyridinyl)morpholine
Precaución: for Research Use Only
Formula:
C₉H₁₁ClN₂O MW: 198.65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00498941 CAS Number: 54231-36-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...