4-Iodo-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

29987-5G 29987-25G
77361-394EA 173.04 USD
77361-394 77373-120
4-Iodo-2-methylbenzoic acid ≥95%
4-Iodo-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MW: 262.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698511
CAS Number: 133232-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR