2-Chloro-6-(trifluoromethyl)nicotinamide ≥97%

Supplier: AOBChem USA

Warning

29944-5G 29944-25G
77361-390EA 79.31 USD
77361-390 77373-118
2-Chloro-6-(trifluoromethyl)nicotinamide ≥97%
2-Chloro-6-(trifluoromethyl)nicotinamide

Precaución: for Research Use Only

Formula: C₇H₄ClF₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD01862659
CAS Number: 386704-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR