Ethyl 2-bromo-3-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

29093-5G 29093-25G
77361-320EA 109.18 USD
77361-320 77373-058
Ethyl 2-bromo-3-fluorobenzoate ≥97%
Ethyl 2-bromo-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD10000923
CAS Number: 1131040-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR