3-Fluorophenyl N,N-diethylcarbamate ≥97%

Supplier: AOBChem USA

Warning

28221-5G 28221-25G
77361-252EA 90.64 USD
77361-252 77373-012
3-Fluorophenyl N,N-diethylcarbamate ≥97%
3-Fluorophenyl N,N-diethylcarbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₄FNO₂
MW: 211.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755848
CAS Number: 205187-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR