2-Bromo-4,5-difluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

27695-5G 27695-25G
77361-214EA 82.4 USD
77361-214 77372-982
2-Bromo-4,5-difluorobenzaldehyde ≥95%
2-Bromo-4,5-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD16659662
CAS Number: 476620-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR