2,6-Dichloro-4-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

27466-5G 27466-25G
77361-204EA 67.98 USD
77361-204 77372-968
2,6-Dichloro-4-methoxypyridine ≥97%
2,6-Dichloro-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD00955755
CAS Number: 17228-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR